Learn More
Cooper Atkins Corporation Thermistor Temperature Instrument
Catalog No. NC0259406
Encompass_Preferred
For those applications where only one temperature is read at a time, the TM99A is an industry standard. It features Min/Max memory, hold, auto shut-off. LCD backlight for dark environments. The TM99A includes a pouch and 1075 general purpose probe.
Catalog No. NC0259406 Supplier Cooper Atkins Corporation Supplier No. TM99A
For those applications where only one temperature is read at a time, the TM99A is an industry standard. It features Min/Max memory, hold, auto shut-off. LCD backlight for dark environments. The TM99A includes a pouch and 1075 general purpose probe.