Learn More
Cole-Parmer Digi-Sense Thermistor Probe, 304 SST, 5 / 32" Dia, 4.5" L, Phono Plug, 10ft Lead
Catalog No. 501961497 Shop All Cole-Parmer Products
Encompass_Preferred
Thermistor probes offer excellent accuracy over the biological temperature range. Probes are electronically isolated and come with a 10 ft. lead with a phono plug (unless otherwise specified). Probes are designed for multiple applications. Time Constant: 1-4 seconds or greater
Catalog No. 50-196-1497 Supplier Cole-Parmer Supplier No. 0849106
Thermistor probes offer excellent accuracy over the biological temperature range. Probes are electronically isolated and come with a 10 ft. lead with a phono plug (unless otherwise specified). Probes are designed for multiple applications. Time Constant: 1-4 seconds or greater