Learn More
Cole-Parmer Traceable Scientific Thermistor Thermometer with Calibration; USB Probe
Catalog No. NC0649825 Shop All Cole-Parmer Products
Encompass_Preferred
This high-precision thermometer is ideal for measuring temperatures in gas systems, reagents, chemical solutions, soil, water baths, ovens, incubators, pharmaceuticals, foods and wastewater. Designed for years of reliable service even in severe environments, the meter features a splash proof membrane keypad. A high-contrast 1/2"-high LCD makes the thermometer easy to read in all types of light. The meter features memory and hold function, internal datalogger that captures the min/max temperature every hour for a 24-hours period, and F/C selection.
Catalog No. NC0649825 Supplier Cole-Parmer Supplier No. 9008009
This high-precision thermometer is ideal for measuring temperatures in gas systems, reagents, chemical solutions, soil, water baths, ovens, incubators, pharmaceuticals, foods and wastewater. Designed for years of reliable service even in severe environments, the meter features a splash proof membrane keypad. A high-contrast 1/2"-high LCD makes the thermometer easy to read in all types of light. The meter features memory and hold function, internal datalogger that captures the min/max temperature every hour for a 24-hours period, and F/C selection.