Learn More
Cole-Parmer TSI AVM430-A CompuFlow Thermoanemometer, manual datalogging
Catalog No. 50633417 Shop All Cole-Parmer Products
Encompass
These thermoanemometers measure air velocity, air volume, and temperature. Display up to three measurements simultaneously on a large backlit LCD. Automatically calculate averages of all your parameters with the push of a button.
Catalog No. 50-633-417 Supplier Cole-Parmer Supplier No. 1025025
These thermoanemometers measure air velocity, air volume, and temperature. Display up to three measurements simultaneously on a large backlit LCD. Automatically calculate averages of all your parameters with the push of a button.