Learn More
Braintree Scientific Thermo Controlled Recovery Pad, (shoe box cage not included)
SDP
Catalog No. 501956034
Encompass
Thermo Controlled Recovery Pad - Animal recovery made simple and easy! Convenient size 13.5" L x 7.5" W, includes3 space gels for continuous heat, 2 space blankets to retain heat, Thermo reflector and turbo heat sink to maximize heat transfer, Non-skid feet for added stability. Fits into a standard 18" L x 9" W. Ultra-Safe Thermo Regulation: will not over heat or burn animal, no electricity used. **Sold for research purposes only and not for human use**
Catalog No. 50-195-6034 Supplier Braintree Scientific Supplier No. TCRP
Thermo Controlled Recovery Pad - Animal recovery made simple and easy! Convenient size 13.5" L x 7.5" W, includes3 space gels for continuous heat, 2 space blankets to retain heat, Thermo reflector and turbo heat sink to maximize heat transfer, Non-skid feet for added stability. Fits into a standard 18" L x 9" W. Ultra-Safe Thermo Regulation: will not over heat or burn animal, no electricity used. **Sold for research purposes only and not for human use**