Learn More
Braintree Scientific Thermo Controlled Surgery Platform w/Space Gel Pads
SDP
Catalog No. 501954216
Encompass
Thermo Controlled Surgery Platform - Easily maintain animal body temperature during surgery! Two flexible LED lights provide direct illumination with no shadows and no heat. Thermo reflector and turbo heat sink to maximize heat transfer. Flex arm, non-obstructive rimless magnifying glass, 2.3x main lens, 4.0x inset lens. Allows continuous eye contact with object when moving into magnification area. Includes Limb restraints, 4 Space Gels for continuous heat. 2 Space Blankets to retain heat and non-skid feet for added stability. Universal input power supply. Size: 13.5" L x 9" W **Sold for research purposes only and not for human use**
Catalog No. 50-195-4216 Supplier Braintree Scientific Supplier No. ASS7T
 Restricted Item
Add to Cart
Add to Cart
Thermo Controlled Surgery Platform - Easily maintain animal body temperature during surgery! Two flexible LED lights provide direct illumination with no shadows and no heat. Thermo reflector and turbo heat sink to maximize heat transfer. Flex arm, non-obstructive rimless magnifying glass, 2.3x main lens, 4.0x inset lens. Allows continuous eye contact with object when moving into magnification area. Includes Limb restraints, 4 Space Gels for continuous heat. 2 Space Blankets to retain heat and non-skid feet for added stability. Universal input power supply. Size: 13.5" L x 9" W **Sold for research purposes only and not for human use**