Learn More
Braintree Scientific Thermo Controlled Surgery Platform w/Deltaphase Pads
SDP
Catalog No. 501954215
Encompass
Thermo Controlled Surgery Platform - Easily maintain animal body temperature during surgery! Two flexible LED lights provide direct illumination with no shadows and no heat. Thermo reflector and turbo heat sink to maximize heat transfer. Flex arm, non-obstructive rimless magnifying glass, 2.3x main lens, 4.0x inset lens. Allows continuous eye contact with object when moving into magnification area. Includes Limb restraints, 1 Deltaphase Pad for continuous heat. 2 Space Blankets to retain heat and non-skid feet for added stability. Universal input power supply. Size: 13.5" L x 9" W **Sold for research purposes only and not for human use**
Catalog No. 50-195-4215 Supplier Braintree Scientific Supplier No. ASS7D
Thermo Controlled Surgery Platform - Easily maintain animal body temperature during surgery! Two flexible LED lights provide direct illumination with no shadows and no heat. Thermo reflector and turbo heat sink to maximize heat transfer. Flex arm, non-obstructive rimless magnifying glass, 2.3x main lens, 4.0x inset lens. Allows continuous eye contact with object when moving into magnification area. Includes Limb restraints, 1 Deltaphase Pad for continuous heat. 2 Space Blankets to retain heat and non-skid feet for added stability. Universal input power supply. Size: 13.5" L x 9" W **Sold for research purposes only and not for human use**