Learn More
Glas Col LLC Thermocouple
SDP
Catalog No. NC0540815
Encompass_Preferred
Thermocouple type "J", recommended range 1-370 degrees C, 24 gauge, 1/8" diameter x 6' long stainless steel probe. 48" long fiberglass indulated duplex lead with stainless steel overbraid with miniature plug. For liquid immersion.Use with Glas-Col controllers and thermometer.
Catalog No. NC0540815 Supplier Glas Col LLC Supplier No. 104A TC105
Thermocouple type "J", recommended range 1-370 degrees C, 24 gauge, 1/8" diameter x 6' long stainless steel probe. 48" long fiberglass indulated duplex lead with stainless steel overbraid with miniature plug. For liquid immersion.Use with Glas-Col controllers and thermometer.