Learn More
Wells Lamont Industrial™ Thermo Cutflex Glove
Catalog No. 18063981 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Medium
Catalog No. Size
18-063-981 Medium
1 options
Catalog No. 18-063-981 Supplier Wells Lamont Industrial™ Supplier No. 135829
Add to Cart
Add to Cart
Thermo Cutflex ANSI Cut Level A7 glove. CFR21 approved for food contact. Made in the USA.
The Whizard™ Thermo CutFlex™ Glove is a single-glove solution, providing exceptional warmth and increased dexterity over existing two-glove solutions.
  • Composite yarn technology with a specialized fiber blend and knit structure provide a thermal barrier to ensure warmth in cold environments.
  • Single-Glove cut resistant seamless knit shell offers more dexterity, improving ergonomics and reducing CTDs.
  • The Thermo CutFlex™ Glove is ideal for cut/processing food applications as well as other cold environments where handling sharp objects require dexterity while maintaining warmth.
Specifications
Construction Seamless Knit
Coating Material Uncoated
Color Hi-vis Orange
Cut Resistance Rating ANSI A7
Hand Ambidextrous
Latex Free Yes
Material Composite Yarn
Product Type Cut-resistant Gloves
Size Medium
Gender Unisex
Glove Cuff Style Elasticated
Lining Unlined
Puncture Rating ASTM F2878 LEVEL 0
Thumb Style Straight