Learn More
MSC Rochester Midland Floor Finish, Dry Bright, Detergent Resistant, ASTM D-2047 Slip Resistant
Catalog No. 502835864
Encompass
This dry bright floor finish provides a brilliant gloss that deepens and becomes tougher with each burnishing. It is ideal for daily maintenance and is recommended for frequent burnishing programs. The product is detergent resistant and meets slip-resistant standard ASTM D-2047.
  • Burnishes to a brilliant gloss "wet look"
  • Becomes more durable with each burnishing
  • Detergent, alcohol, and scuff resistant
  • Meets slip resistant standard ASTM D-2047
  • Features excellent leveling characteristics
  • Easy to maintain
  • Provides excellent gloss retention
  • Recommended for frequent burnishing programs
  • Covers up to 2,000 sq ft per gallon
  • Has a mild fragrance and opaque white color
  • Features a pH of 9.0
Catalog No. 50-283-5864 Supplier MSC Supplier No. 81212284
This dry bright floor finish provides a brilliant gloss that deepens and becomes tougher with each burnishing. It is ideal for daily maintenance and is recommended for frequent burnishing programs. The product is detergent resistant and meets slip-resistant standard ASTM D-2047.
  • Burnishes to a brilliant gloss "wet look"
  • Becomes more durable with each burnishing
  • Detergent, alcohol, and scuff resistant
  • Meets slip resistant standard ASTM D-2047
  • Features excellent leveling characteristics
  • Easy to maintain
  • Provides excellent gloss retention
  • Recommended for frequent burnishing programs
  • Covers up to 2,000 sq ft per gallon
  • Has a mild fragrance and opaque white color
  • Features a pH of 9.0