Learn More
Cole-Parmer Extech 445703 Big Digit Thermohygrometer
Supplier:  Cole-Parmer 3780210
Encompass
Designed to monitor relative humidity, and temperature. Features include a super large display, C/F switchable, Min/Max with reset function, low battery indication and a built-in stand for sitting on a desk and hardware for mounting on a wall.
Catalog No. 50-993-028