Learn More
Thermo Scientific™ 96-Well Microtiter Microplates
GSA_VAGREENER_CHOICE
Catalog No. 14245146 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
14-245-146 Clear
1 options
Catalog No. 14-245-146 Supplier Thermo Scientific™ Supplier No. 230511
  • $343.00
    $250.65 / Case of 50
    Save $92.35
    27% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Allow miniaturization of assays and other applications including high-volume DNA library manipulation and solid-phase binding
  • For high-throughput screening and research
  • Clear plates for colorimetric applications
  • Standard footprint, 128 × 86mm
  • Available as both polystyrene square wells and vinyl square wells
  • Vinyl Microtiter Microplates are used for complement fixation, agglutination, and blood typing
  • Styrene Microtiter Microplates are used for complement fixation, agglutination, microbiology testing, and blood typing
Order Info
Polystyrene microplates are available as 50 units/package. Vinyl microplates are available as 25 or 100 units/package, depending on product.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 8 x 12
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Non-treated
Well Shape Round Bottom (U-well)
Quantity 50