Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Thermo Scientific™ 96-well PCR Base 

Catalog No. A55330 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
A55330 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A55330 Supplier Thermo Scientific™ Supplier No. A55330
Add to Cart
Add to Cart
This item is not returnable. View return policy

Pipette with confidence using the specifically designed Thermo Scientific 96-well PCR Base in the SureTect workflow.

Easily and quickly carry out the lysis and PCR setup during the SureTect workflow using the Thermo Scientific™ 96-well PCR Base to securely hold the lysis and assay tubes. Fitting perfectly with the specifically designed Applied Biosystems™ QuantStudio™ 5 and Applied Biosystems™ SimpliAmp™ trays, the 96 well PCR Base allows confident pipetting and easy transference of tubes from rack to instrument in a single step.

Designed specifically for use with the Thermo Scientific™ SureTect™ assays, the 96-well PCR Base is one in a suite of tools enabling a simple, streamlined workflow for maximum efficiency and productivity in your laboratory.

Use the Thermo Scientific 96-well PCR Base to securely hold the lysis and assay trays and tubes when pipetting into the SureTect lysis and PCR tubes enabling a fast, easy to use workflow

Quick and Easy

Use the Thermo Scientific 96-well PCR Base to give confidence when handling and pipetting into the lysis and PCR tubes during the SureTect workflow. This tool is designed specifically for holding the SureTect tube strips during pipetting and along with the SimpliAmp and Quant Studio 5 trays enables the secure and easy handling and transferring of the SureTect lysis and assay tubes through the workflow. The rugged material and design endures constant use so can be used time and time again.

Combined with our comprehensive portfolio of assays and world-class instrumentation you can perform food safety tests confidently knowing exactly what you are getting – quality combined with simplicity.

We are committed to enabling you to experience the true confidence in your results, right-first time, every time, and the Thermo Scientific™ SureTect™ Real-Time PCR System is continually evolving to support your laboratory and testing needs.

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Color Black
Product Type 96-well PCR Base
For Use With (Equipment) Applied Biosystems™ QuantStudio™ 5 and Applied Biosystems™ SimpliAmp™ trays
For Use With (Application) SureTect workflow
Quantity 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.