Description
- 96-well plates contain two 0.25mm holes laser cut at conical ends of each well
- Purifies full-length proteins from SDS-PAGE gels
- MALDI-TOF compatible
- Protocol can be followed by trypsin digest if required
Specifications
Specifications
| Array | 8 x 12 |
| No. of Wells | 96 |