Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ AminoLink™ Reductant

Catalog No. PI44892 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 x 1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI44892 2 x 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI44892 Supplier Thermo Scientific™ Supplier No. 44892
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Kit contains pre-packed AminoLink Columns, coupling buffer, quenching buffer, wash solution, sodium cyanoborohydride solution, and column accessories

Easily immobilize antibodies and other proteins for use in affinity purification

AminoLink Coupling Resin is crosslinked 4% beaded agarose that has been activated with aldehyde groups. Proteins and other molecules with primary amines can be covalently attached (immobilized) to the resin to make chromatography columns for use in affinity purification. The aldehyde groups form stable secondary amine bonds with primary amines such as exist in the side chain of lysine (K) residues, which are generally abundant and readily accessible in proteins. Once a protein is immobilized, the prepared affinity resin can be used for a variety of batch and column affinity purification methods involving binding interactions with the immobilized protein. The resin and linkage are stable in most binding and elution conditions typically used in affinity chromatography, enabling prepared resin to be used for multiple rounds of affinity purification procedures.

Highlights:
AminoLink Coupling Resin – aldehyde-activated crosslinked 4% beaded agarose
Ideal for antibodies and other proteins – immobilize molecules via primary amines (-NH2)
Flexible coupling conditions – efficient (>85%) coupling over a wide range of pH (4 to 10) and buffer conditions (PBS or other non-amine buffer with or without organic solvent
Stable, permanent immobilization – coupling reaction results in stable, leak-resistant secondary amine bond between resin and ligand
Better than immobilization to CNBr-activated agarose – bond is more stable and uncharged, resulting in less nonspecific binding in affinity purification procedures
Versatile and reusable – prepared affinity resin is adaptable to column and batch affinity techniques and the resin is reusable for typical applications based on protein binding interactions

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store desiccated at room temperature. Shipped at ambient temperature
Form Powder
Product Line AminoLink
Quantity 2 x 1 g
Type Resin
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.