Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ AMS-4 Continuous Beta Air Monitors
GSA_VA

Catalog No. p-3683460 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Iodine Type 508 Filter Paper
Particulate Type LB5211 Filter Paper
Serial Communication Cable
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-175-532 Particulate Type LB5211 Filter Paper
19-175-533 Iodine Type 508 Filter Paper
19-175-535 Serial Communication Cable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-175-532 Supplier Thermo Scientific™ Supplier No. FIFP10
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Provides early warning to workers exposed to potential airborne releases of beta emitting particulates; lightweight and robust design accommodates both fixed and portable use applications

Continuous air monitor (CAM) offering beta particulate, radioiodine, or noble gas detection.

  • Detachable sampling head
  • Reports both DAC and cpm
  • Real time gamma background subtraction
  • Stand alone or network configuration
  • Mass flow measurements
  • Fast and slow concentration alarms, cpm alarm, DAC-hour alarm
  • Computer calibration
  • Can also be used for stack monitoring of effluent releases using the in-line detector head

  • Keypad: 18 keys, full numeric plus function keys
  • Status Indicators: Front panel lights display READY and MALFUNCTION conditions, red alarm strobe light, sonalert
  • Printer: RS-232 serial DB9
  • Data Log Buffer: 2000 entries of combined Concentration and Status
  • Change information: Equivalent to 83 days of 1 hour detail, nearly 14 days of 10 mintes detail or over 24 hours of 1 minute detail
  • Alarm/Fail Relays: SPST, 15A at 30VDC/120VAC resistive
  • Analog Output: 0 to 5VDC output, five decade logarithmic

Compliance

Meets Regulatory Guide 8.25 and revised 10 CFR 20 criteria

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Filter paper for the AMS-4; 0.47 mm, particulate type LB5211 or equivalent
Product Type Particulate Type LB5211 Filter Paper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.