Learn More
Description
The SoftFit-L System is compatible with Rainin LTS pipettes made by Rainin Instrument LCC.
- Available in low retention
- Sterile tips are certified free from RNase, DNase and pyrogen
- Manufactured according to the industry's highest quality standards
Specifications
Specifications
| Volume (Metric) | 500 μL |
| Sterility | Non-sterile |
| Format | Bulk |
| For Use With (Equipment) | Rainin LTS LiteTouch |
| Product Line | ART |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.