Learn More
Thermo Scientific™ Barnstead™ Deionization System
GSA_VA
Catalog No. p-169304
Change view
Click to view available options
Type:
Deionization Cartridge
Deionization System
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
09-050-267 Deionization Cartridge
09-034-175K Deionization System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-050-267 Supplier Thermo Scientific™ Supplier No. D5021
Only null left
Add to Cart
Add to Cart
For the Nanopure, E-pure Water Systems
Deionization System
For the Nanopure, E-pure Water Systems
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Pore Size 60 μm
Resin Type Mixed Bed
For Use With (Application) Deionization System
For Use With (Equipment) Quad, E-Pure, Deionization Cartridge
Type Deionization Cartridge