Learn More
Thermo Scientific™ Barnstead™ Easypure™ Ultrapure Cartridge
Catalog No. 19025350 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Replacement Ultrafilter
Ultrapure Cartridge
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
19-025-350 Replacement Ultrafilter
09-050-46 Ultrapure Cartridge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-025-350 Supplier Thermo Scientific™ Supplier No. FL550X1
Add to Cart
Add to Cart
For use with Thermo Scientific Barnstead Easypure ultrapure water purification systems
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Nanopure R, Infinity and EasyPure™ water purification system
Model FL550X1
Type Replacement Ultrafilter