Learn More
Thermo Scientific™ Barnstead™ Pump Protectors for Water Purification Systems
Shop All Thermo Scientific Products
Description
- For installation in a pressurized feedwater line
- Shuts off pump if feedwater pressure falls below 5psi (0.35kg/cm2)
- Plugs into Nanopure (Nanopure Quad-style, Nanopure II or older Nanopure), E-pure, or older Easypure UV, UF, UV/UF and LF systems
- Can also be used with select Nanopure DIamond™ systems
- Includes everything necessary for installation
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| For Use With (Equipment) | E-Pure Ultrapure Water Purification System |