Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Barnstead Pura-Lite Water Purity Monitor

Catalog No. 0903429 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
B-pure Water System
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
09-034-29 B-pure Water System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-034-29 Supplier Thermo Scientific™ Supplier No. E3450
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with B-Pure Water System

  • Optional Pura-Lite indicator light
  • For use with the B-Pure water system
  • Light illuminates at 50,000 ohm-cm
  • Voltage: 120V
  • Shipping weight: 10.4 ounces

TRUSTED_SUSTAINABILITY

Specifications

Type Water Purity Indicator
Voltage 110 V
For Use With (Equipment) B-pure Water System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.