Learn More
Thermo Scientific™ Barnstead™ Water Purification System Replacement Cartridges
Catalog No. 09050247
Change view
Click to view available options
Type:
Cartridge
Organicfree Cartridge
Pretreatment Cartridge
For Use With (Equipment):
Easypure Reverse Osmosis System
Nanopure Infinity System
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
09-050-247 Cartridge Nanopure Infinity System
09-050-67 Pretreatment Cartridge Easypure Reverse Osmosis System
09-050-246 Organicfree Cartridge Nanopure Infinity System
09-050-226 Pretreatment Cartridge Nanopure Infinity System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 09-050-247 Supplier Thermo Scientific™ Supplier No. D50253
For Easypure™ RO water purification system
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Water Purification System Replacement Cartridge
For Use With (Equipment) Nanopure Infinity System
Type Cartridge