Learn More
Thermo Scientific™ BarnsteadPUREpack™ Cartridges
GSA_VA
Catalog No. 0902531
Change view
Click to view available options
Type:
Filter Replacement Cartridge
Purepack 1 with Filter Replacement Cartridge
Purepack 2 Replacement Cartridge
Purepack Reverse Osmosis Replacement Cartridge
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
09-025-31 Purepack 1 with Filter Replacement Cartridge
09-025-32 Filter Replacement Cartridge
09-025-34 Purepack 2 Replacement Cartridge
09-025-35 Purepack Reverse Osmosis Replacement Cartridge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 09-025-31 Supplier Thermo Scientific™ Supplier No. D50265
Add to Cart
Add to Cart
Choose from an array of Thermo Scientific™ Barnstead™ PUREpack™ Cartridges for select Thermo Scientific™ Barnstead™ water systems, based on your application.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Capacity 1121 grains
Feedwater Source Reverse Osmosies, Distilled, Ultrafiltered
Pore Size 0.2 μm
For Use With (Application) Reverse osmosis, distillation, ultrafiltration feedwater source
Type Purepack 1 with Filter Replacement Cartridge