Learn More
Thermo Scientific™ Chemitest Temperature Indicator Strips for Steam Sterilizers
GSA_VA
Catalog No. 1449027 Shop All Thermo Scientific Products
Change view
Click to view available options
Chemitest Indicator Strips
Catalog No.
14-490-27 Chemitest Indicator Strips
1 options
Catalog No. 14-490-27 Supplier Thermo Scientific™ Supplier No. AY759X2
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Chemical Indicator Strips 132 to 135°C (270 to 275°F)
Temperature verification strips verify that a sterilizer has reached the proper temperature through a color change. Daily verification ensures that items being processed have been run through a cycle, preventing the accidental use of items that have not been adequately processed.
Chemitest™ Temperature Indicators
  • Verify that sterilizer reached adequate sterilization temperature during a cycle
  • Can also be used to verify that an item has been run through a sterilization cycle
  • Available in pouches, tabs, strips, and tape
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Type Biological Indicator
For Use With (Application) Steam sterilization
Quantity 250 vials/box
Size L x W: 3.38 x 4.38 in.
Sterilization Indicator Yes