Learn More
Thermo Scientific™ Capit-All Flex Automated Decapper
Catalog No. 14377900 Shop All Thermo Scientific Products
Change view
Click to view available options
Decapping Format:
96-Channel
Catalog No. Decapping Format
14-377-900 96-Channel
1 options
Catalog No. 14-377-900 Supplier Thermo Scientific™ Supplier No. 4120FLEX
Add to Cart
Add to Cart
Request a Quote
The Thermo Scientific™ Capit-All™ Flex Automated Decapper is a full-service, automated capping and decapping solution for 48- and 96-tube racks.
Experience unparalleled efficiency with our advanced cryotube decapper system, designed to meet the needs of modern laboratories. Compatible with both Matrix and Nunc cryotubes in 96 and 48 formats, this all-in-one instrument combines the functionality of four separate instruments, significantly reducing your laboratory's footprint and costs. Switch seamlessly between tube types without the hassle of adapters or part changes. The large 7 in. angled touchscreen, operable with or without gloves, ensures ease of use and visibility. A built-in drip tray acts as a barrier between caps and tubes to prevent cross-contamination and maintain the integrity of your samples. Our instrument includes tube verification to minimize errors and supports both full and partial racks, adapting to your specific needs. Whether used on the benchtop or an integrated system, it offers flexibility for your storage applications.
  • Compatible with both Matrix and Nunc cryotubes in 96 and 48 formats
  • Functionality of 4 instruments in one, reducing footprint and spend
  • Switch between tube types without the need of adapters or changing parts
  • Large angled 7 in. touchscreen that can be used with or without gloves
  • Built-in drip tray that acts as a barrier between caps and tubes to help prevent cross-contamination and maintain sample integrity
  • Tube verification to prevent errors
  • Works with full or partial racks of tubes
  • Can be used for benchtop or integrated applications - fits inside a Biosafety Cabinet
TRUSTED_SUSTAINABILITY
Specifications
Compatible Storage Tubes Matrix and Nunc tubes in 96 and 48 format
Decapping Format 96-Channel
Description Automated Decapper
Operation Touchscreen User Interface
Voltage 100/240 VAC
Positions Ω