Learn More
Fisherbrand™ Castaloy™ Chain Clamp
Catalog No. 05745Q Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
05-754 Q Clamp Holder
Catalog No. For Use With (Equipment)
05-745Q 05-754 Q Clamp Holder
1 options
This item is not returnable. View return policy
Catalog No. 05-745Q
This item is not returnable. View return policy
For holding bottles, beakers and other large objects
For holding bottles, beakers and other large objects
  • Holds 76 to 165mm (3 to 6.5 in.) bottles and large objects
  • Fast, sure way to secure large equipment to rods and lattices
  • Loop steel chain around object; attach to hook on movable slide; tighten slide with large, nonslip knob
  • Holds equipment firmly in place at any angle, variable distance from support
  • Arm is 15cm (6 in.) long and 13mm (0.5 in.) diameter
  • Use with 05-754Q clamp holder
Specifications
Quantity 1 Ea.
Type Chain Clamp
Material Stainless Steel
For Use With (Equipment) 05-754 Q Clamp Holder
For Use With (Application) For holding bottles, beakers and other large objects
Length (English) Arm 6 in.
Length (Metric) Arm 15.24 cm