Learn More
Fisherbrand™ Castaloy™ Clamp Swivel Holder
Catalog No. 05762Q Shop All Fisherbrand Products
Change view
Click to view available options
Jaw Type:
Swivel
Catalog No. Jaw Type
05-762Q Swivel
1 options
This item is not returnable. View return policy
Catalog No. 05-762Q
This item is not returnable. View return policy
Tilt clamp at any angle in parallel planes
Tilt clamp at any angle in parallel planes
  • Holder will not fall apart when setscrew is loosened to permit one end to swivel
  • With 45° angle thumbscrews
  • Grips rods up to 0.75 in. (19mm) diameter
Specifications
Quantity 1 Ea.
Type Swivel Clamp Holder
Includes 45° Angle thumbscrews
Jaw Type Swivel