Learn More
Fisherbrand™ Castaloy™ Hosecock Clamp
Catalog No. 05847Q Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Accurate Regulation of Flow
Catalog No. For Use With (Equipment)
05-847Q Accurate Regulation of Flow
1 options
Catalog No. 05-847Q
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product
Fine-machined adjustment screw for very accurate regulation of flow
Fine-machined adjustment screw for very accurate regulation of flow
  • Threaded keeper screw on side of clamp can be raised to allow easy positioning of tubing, then screwed back into base of clamp
  • For tubing up to 0.75 in. (19mm) O.D.
  • Concave lower bearing surface, convex upper which reduces amount of pressure required for complete closure
Specifications
Quantity 1 Ea.
Type Hosecock Clamp
Includes Adjustment screw
For Use With (Equipment) Accurate Regulation of Flow
For Use With (Application) For accurate regulation of flow
Size (Metric) Grip 19 mm O.D.
Size (English) Grip 0.75 in. O.D.