Learn More
Unity Lab Services™ Thermo Scientific Centrifugation Validation and Temperature Map Documention for Superspeed and Ultra centrifuges: Thermo Scientific, Sorvall
Catalog No. iopqde03493
Catalog No. IOPQDE03493 Supplier Unity Lab Services™ Supplier No. IOPQDOCE89003493
Only null left
Add to Cart
Add to Cart
Original equipment manufacturer service provider for Thermo Fisher Scientific. All protocols designed to optimize efficiency are included with each Thermo Scientific Centrifugation Validation and Cycle Testing Documention for Superspeed and Ultra centrifuges: Thermo Scientific, Sorvall
IQ/OQ/Cycle Testing documentation that supports regulatory requirements and industry standards for instrument compliance. Our documents are kept up to date by conducting annual reviews. We work directly with our product experts to have the most updated documentation available.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Thermo Scientific Centrifugation including Superspeed and Ultra centrifuges: Thermo Scientific, Sorvall
Description Documentation Only for the following, Services Not included: Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, perform min/max speed, brief/extended time and temperature testing (for refrigerated models). Cycle Testing: Point-In-Time test exercise of pre-defined duration that surpasses singular calibration and routine speed, time, and temperature monitoring activities.
Location Contact your local sales representative for a list of standard and extended coverage areas.
Includes IQ/OQ/Cycle Testing Documentation