Learn More
Unity Lab Services™ Thermo Scientific™ CO2 Incubators Validation Service for Thermo Scientific™ CO2 incubators
Shop All Thermo Scientific ProductsDescription
IQ
- Unpacking, Assembly and Installation Verifies that the equipment, manuals, supplies and any other accessories arrived undamaged as specified in the sales order. Verifies equipment is assembled and installed and any initial diagnostics tests have been performed. Any abnormal event(s) observed during assembly and installation have been documented.
- Utility, Facility and Environment Verifies that the installation site satisfactorily meets manufacturer-specified environmental requirements.
OQ
- System Component Information Detailed configuration information for each system component have been recorded.
- OQ Limits: Record a list of manufacturer recommended limits for ensuring that the system is operating as expected.
- Equipment Operational Tests and Results: Test important equipment functions to verify that the equipment operates as intended by the manufacturer and required by the user. This is a group of selected important equipment parameters for testing according to the intended use of the equipment.
Key Tests: Door Ajar, Power Failure, CO2 Concentration and Temperature levels
Specifications
Specifications
| For Use With (Equipment) | Thermo Scientific CO2 Incubators |
| Product Type | IQ/OQ Service |
| Location | Contact your local sales representative for a list of standard and extended coverage areas. |
| Includes | Travel to major metropolitan areas. Does not include Installation. Instrument must be installed, powered-up and set temperature point for 24 hours before on-site service |