Learn More
Unity Lab Services™ Thermo Scientific™ Combination Refrigerator/Freezer Validation Service
Catalog No. 11675247 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Thermo Scientific Combination Refrigerator/Freezers
Catalog No. For Use With (Equipment)
11-675-247 Thermo Scientific Combination Refrigerator/Freezers
1 options
Catalog No. 11-675-247 Supplier Unity Lab Services™ Supplier No. IOQPCKE89003679
Add to Cart
Add to Cart
Stay compliant with our Industry-Exclusive, No-Charge Requalification Guarantee included with each Thermo Scientific Combination Refrigerator/Freezer Validation Service for Jewett combination Refrigerator/Freezer
IQ
  • Unpacking, Assembly and Installation Verifies that the equipment, manuals, supplies and any other accessories arrived undamaged as specified in the sales order. Verifies equipment is assembled and installed and any initial diagnostics tests have been performed. Any abnormal event(s) observed during assembly and installation have been documented.
  • Utility, Facility and Environment Verifies that the installation site satisfactorily meets manufacturer-specified environmental requirements.
OQ System Component Information Detailed configuration information for each system component have been recorded.
  • OQ Limits: Record a list of manufacturer recommended limits for ensuring that the system is operating as expected.
  • Equipment Operational Tests and Results: Test important equipment functions to verify that the equipment operates as intended by the manufacturer and required by the user. This is a group of selected important equipment parameters for testing according to the intended use of the equipment.
- Key Tests: Door Ajar, Power Failure, Temperature Alerts
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Thermo Scientific Combination Refrigerator/Freezers
Description Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, open door and power loss testing.
Product Type IQ/OQ Service
Location Contact your local sales representative for a list of standard and extended coverage areas.
Includes Travel to major metropolitan areas. Does not include Installation. Instrument must be installed, powered-up and set temperature point for 24 hours before on-site service