Learn More
Thermo Scientific™ CryoExtra™ Racks and Accessories
Specifications
Specifications
| Type | Cryo Mini Rack |
| For Use With (Equipment) | Thermo Scientific CryoExtra 20, 40, 80 and 94 Rack |
| Includes | 2mL, 13 Cardboard boxes (25 cell divider) |
| Holds | 2mL, 13 Cardboard Boxes (25-cell Divider) |