Description
Sample thermocouple sensors
Thermocouple sensors for cryo tubes, sheath straw and vial samples.
- Include male plug
For controlled-rate freezing applications
Specifications
Specifications
| Type | Ribbon Type |
| For Use With (Equipment) | Cyrogenic Freezer |
| Plug Type | Male |
| Holds | Cryo Tube, Sheath Straw, Vial |