Learn More
Thermo Scientific™ DNA Quantitation Standards
Description
Includes
Standards supplied in 50μg/mL ReddyRun™ Buffer include a separate vial of 10X ReddyRun Buffer to add to samples.
- Offer a choice of fragment sizes
- Concentration of each standard: 100μg/mL
- Provide versatile quantitation for ethidium bromide stained gels
- Mirror the effect seen in samples—larger fragments tend to form sharper bands and smaller fragments more diffuse bands in agarose gels
- Supplied in TE storage buffer
Fragment sizes:
QSK-101, QSK-101L: 200 bp, 500 bp, and 1000 bp plus two prelabeled tubes
QS1000, QS1000LD: 1000bp
QS200, QS200LD: 2000bp
QS500LD: 500bp
Specifications
Specifications
| Quantity | 250/Cs. |
| Product Type | Dilution and Delivery Device System |