Learn More
Description
Includes
Standards supplied in 50μg/mL ReddyRun™ Buffer include a separate vial of 10X ReddyRun Buffer to add to samples.
- Offer a choice of fragment sizes
- Concentration of each standard: 100μg/mL
- Provide versatile quantitation for ethidium bromide stained gels
- Mirror the effect seen in samples—larger fragments tend to form sharper bands and smaller fragments more diffuse bands in agarose gels
- Supplied in TE storage buffer
Fragment sizes:
QSK-101, QSK-101L: 200 bp, 500 bp, and 1000 bp plus two prelabeled tubes
QS1000, QS1000LD: 1000bp
QS200, QS200LD: 2000bp
QS500LD: 500bp
Specifications
Specifications
| Concentration | Standards: 100 μg/mL, Buffer: 50 μg/mL, Separate vial of 10X ReddyRun Buffer to add to samples. |
| Form | Liquid |
| Product Type | DNA Molecular Weight Marker |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.