Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DyLight™ 650-4xPEG NHS Ester

Catalog No. PI62274 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI62274 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI62274 Supplier Thermo Scientific™ Supplier No. 62274
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

PEGylated far-red dye for enhanced fluorescence and reduced non-specific binding.

Thermo Scientific DyLight 650-4xPEG Amine-Reactive Dye is a derivative of our high-performance DyLight 650 Dye that can be used to fluorescently label antibodies and other proteins for far-red detection.

The DyLight 680-4xPEG dye contains 4 polyethylene glycol (PEG) chains that are non-cytotoxic, enhance fluorescence and reduce nonspecific binding of conjugates made with them. Conjugates made with DyLight 680-4xPEG Dye can be used as molecular probes for cellular imaging and other fluorescence detection methods. The far-red to NIR fluorescence properties of DyLight 680-4xPEG Dye make it especially useful in a variety of biological, chemical, and pharmaceutical applications, including in vivo imaging. The PEG chains also improve solubility of the dyes and labeled molecules in aqueous solution, aid in cell permeability and improve tissue retention.

Highlights:

  • High fluorescence intensity - significantly brighter fluorescence than Alexa Fluor™ 647
  • PEGylated - improves solubility in aqueous solution and aids in cell permeability
Applications:
  • Fluorescence microscopy
  • In vivo or ex vivo imaging
  • Cell-based assays
  • Flow cytometry/fluorescence-activated cell sorting (FACS)
Product Details:
DyLight 650-4xPEG Amine-Reactive Dye is activated with an N-hydroxysuccinimide (NHS) ester moiety to react with exposed N-terminal α-amino groups or the ε-amino groups of lysine residues to form stable amide bonds.
Typical labeling reactions require the dye to first be dissolved in anhydrous dimethyl formamide (DMF) or another suitable organic solvent before adding a specific molar amount of dye to an amine-free buffer containing the protein to be labeled. However, the high solubility of DyLight Fluors permits protein solutions to be added directly to the labeling reagent.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Product Type Dye
Solubility DMF (Dimethylformamide)
Content And Storage Store desiccated at -20°C.
Chemical Reactivity Amine
Reactive Moiety Active Ester, Succinimidyl Ester, NHS Ester
Label or Dye DyLight™ 650
Label Type DyLight Fluors
Product Line DyLight
Emission 672
Excitation 652
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.