Learn More
Thermo Scientific™ Evolution™ One, OnePlus and Pro Spectrophotometer Solid Sample Holders
Catalog No. p-3522023
Change view
Click to view available options
Holds:
Std 2 x 3 in. Sample Holders
Type:
Solid Sample Holder
Catalog No. Holds Type
14-387-503 Std 2 x 3 in. Sample Holders Solid Sample Holder
1 options
Catalog No. 14-387-503 Supplier Thermo Scientific™ Supplier No. 222217200
Only null left
Add to Cart
Add to Cart
Designed to hold solid samples for transmittance measurements
Solid Sample Holder
  • Provides easy mounting of solid samples like filters, plastic, glass and optics
  • Accommodates standard 2 x 3 in. sample holders
Universal Sample Holder
  • Holds round or square optical samples; glass or plastic samples; lenses from sunglasses or goggles
  • Requires one Solid Sample Holder (14-387-503)
TRUSTED_SUSTAINABILITY
Specifications
Type Solid Sample Holder
For Use With (Equipment) Thermo Scientific™ Evolution™ One, OnePlus and Pro UV-visible spectrophotometers
Holds Std 2 x 3 in. Sample Holders