Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ EZ-Link™ NHS-Desthiobiotin

Catalog No. PI16129 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI16129 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI16129 Supplier Thermo Scientific™ Supplier No. 16129
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Label intracellular proteins for affinity purification with streptavidin beads

Thermo Scientific EZ-Link NHS-Desthiobiotin is a short, membrane permeable, amine-reactive labeling reagent whose biotin-like group is elutable from streptavidin, making it ideal for intracellular protein labeling and purification.

NHS-Desthiobiotin is a variant of biotin that is activated as an NHS ester to covalently label primary amines (-NH2) of proteins or other molecules with desthiobiotin groups. The desthiobiotin tag binds to streptavidin and other biotin-binding proteins with high specificity yet readily elutes with mild conditions (i.e., by competitive displacement with regular, free biotin). As such, this reagent is a useful alternative to NHS-Biotin (Part No. 20217) for avidin-biotin techniques in which nondenaturing elution of the labeled proteins is desired. This simplest and smallest variety of amine-reactive desthiobiotin reagent is uncharged and cell membrane permeable; therefore, NHS-Desthiobiotin can be used with intact cells for intracellular protein labeling.

Highlights:

  • Desthiobiotin - analog of biotin allows easy elution from streptavidin, an ideal feature for affinity purification applications
  • Protein labeling - tag antibodies or proteins in purified or mixed samples to enable high recovery in pull-down assays with streptavidin beads
  • Membrane-permeable - achieve intracellular labeling with intact cells because the small, uncharged reagent easily diffuses across cell membranes
  • Amine-reactive - reacts with primary amines (-NH2), such as the side-chain of lysines (K) or the amino-termini of polypeptides
Desthiobiotin vs. Biotin:
Desthiobiotin is a single-ring, sulfur-free analog of biotin that binds to streptavidin with nearly equal specificity but less affinity than biotin (1/Kd = 10^11 vs. 10^15M, respectively)[1-2]. Consequently, desthiobiotinylated bait proteins and their interacting partners can be eluted readily and specifically from streptavidin affinity resin using mild conditions based on competitive displacement with free biotin. For pull-down assay experiments with biological samples, this soft-release characteristic of desthiobiotin also helps to minimize co-purification of endogenous biotinylated molecules, which remain bound to streptavidin upon elution of the target protein complexes with free biotin. The modified avidin-biotin affinity system also eliminates the need to use harsh elution conditions that might disassociate complexes and/or damage the target protein or cell. Desthiobiotin-based techniques are ideal when using native or recombinant proteins that are not expressed with a fusion tag and when isolating captured proteins under native conditions, such as targeting intact cells or cell surface proteins.
Labeling with NHS Ester Reagents:
N-Hydroxysulfosuccinimide (NHS) esters of biotin are the most popular type of biotinylation reagent. NHS-activated biotins react efficiently with primary amino groups (-NH2) in alkaline buffers to form stable amide bonds. Proteins typically have several primary amines that are available as targets for labeling, including the side chain of lysine (K) residues and the N-terminus of each polypeptide. Most sulfo-NHS esters are directly water soluble but not membrane permeable. Plain NHS esters are less soluble in aqueous buffers but are membrane permeable.

TRUSTED_SUSTAINABILITY

Specifications

Product Type NHS-Desthiobiotin
Quantity 50 mg
Content And Storage Store at -20°C.
Product Line EZ-Link
Cell Permeability Cell-Permeant
Chemical Reactivity Amine
Reactive Moiety Active Ester, Succinimidyl Ester, NHS Ester
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide)
Label or Dye Biotin Analog
Spacer Short
Label Type Biotin & Analogs
Show More Show Less
Can you provide the shelf-life for EZ-Link NHS-Desthiobiotin?

EZ-Link NHS-Desthiobiotin is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (-20 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.