Learn More
Thermo Scientific™ Buffers for MagJET™ Blood gDNA Kit
Catalog No. FERR2371 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
40 mL
60 mL
90 mL
100 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR2371 40 mL
FERR2352 90 mL
FERR2342 60 mL
FERR2361 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. FERR2371 Supplier Thermo Scientific™ Supplier No. R2371
Wash lysed samples to remove any contaminants or inhibitors from the isolated genomic DNA. Thermo Scientific™ MagJET™ Wash Buffer 2 (concentrated) is a component of the MagJET™ Whole Blood gDNA Kits (K2741/2742) and may be purchased separately in 40mL aliquots.
The Thermo Scientific™ MagJET™ Whole Blood Genomic DNA Kit is designed for fast and efficient purification of genomic DNA from fresh or frozen whole blood treated with EDTA or citrate, as well as blood fractions. The kit utilizes paramagnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) MagJET Plant gDNA Kit
Quantity 40 mL