Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Buffers for MagJET™ gDNA Kit

Catalog No. FERR2471 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
35 mL
50 mL
135 mL
Product Type:
Concentrated Wash Buffer 2
Lysis Buffer
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
FERR2471 50 mL Concentrated Wash Buffer 2
FERR2442 135 mL Lysis Buffer
FERR2441 35 mL Lysis Buffer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. FERR2471 Supplier Thermo Scientific™ Supplier No. R2471

missing translation for 'validMainframeMsg'

The Thermo Scientific™ MagJET™ Genomic DNA Kit is designed for fast and efficient purification of genomic DNA from tissue and cell cultures, bacteria and yeast, as well as from human body samples, such as buccal and urogenital swabs, urine, saliva and hair follicles. The kit utilizes paramagnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) MagJET gDNA Kit
Product Type Concentrated Wash Buffer 2
Quantity 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.