Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lysis Buffer B for MagJET Plant gDNA Kit

Catalog No. FERR2491 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
8.4 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR2491 8.4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERR2491 Supplier Thermo Scientific™ Supplier No. R2491

missing translation for 'validMainframeMsg'

The Thermo Scientific™ MagJET™ Plant Genomic DNA Kit is designed for fast and efficient purification of genomic DNA from a variety of plant species and tissues without phenol/chloroform extraction.

The Thermo Scientific™ MagJET™ Plant Genomic DNA Kit is designed for fast and efficient purification of genomic DNA from a variety of plant species and tissues without phenol/chloroform extraction. The kit utilizes magnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.

TRUSTED_SUSTAINABILITY

Specifications

Buffer Lysis Buffer
Format Kit
Product Line MagJET
Product Type Lysis Buffer
Quantity 8.4 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.