Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MagJET NGS Cleanup and Size Selection Kit

Catalog No. p-4861474
Change view
Click to view available options
Quantity:
1 x 96 Preps
10 Preps
10 x 96 Preps
4 x 96 Preps
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERK2822 4 x 96 Preps
FERK2828 10 Preps
FERK2821 1 x 96 Preps
FERK2823 10 x 96 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. FERK2822 Supplier Thermo Scientific™ Supplier No. K2822

Thermo Scientific MagJET NGS Cleanup and Size Selection kit is designed for efficient and robust DNA fragment library cleanup and size-selection from a variety of enzymatic reaction mixtures including PCR, ligation, adapter addition and DNA end-repair reaction mixes.

Thermo Scientific MagJET NGS Cleanup and Size Selection kit is designed for efficient and robust DNA fragment library cleanup and size-selection from a variety of enzymatic reaction mixtures including PCR, ligation, adapter addition and DNA end-repair reaction mixes. The kit utilizes high-capacity MagJET paramagnetic bead technology that ensures efficient recovery of DNA fragment library within a desired fragment length range. The purified DNA fragments are free of any next-generation sequencing workflow inhibiting components, such as sequencing adapters, primer dimers, unincorporated nucleotides, enzymes or salts, and are ready-to-use in downstream NGS applications. No prior DNA fragment library purification is required before the cleanup and size-selection procedures.

The kit is suitable for cleanup or size selection of 5 ng to 5 μg of DNA fragment library. MagJET cleanup and adapter removal protocols can easily be automated for higher throughput on automatic platforms such as Thermo Scientific KingFisher Flex and Thermo Scientific KingFisher Duo.

Highlights
Robust—reliable cleanup and size selection from different reaction mixtures and buffers
Flexible—user-defined size-selection of exact DNA fragment range
NGS-grade purity—complete removal of NGS adapters, primers, nucleotides, enzymes and other reaction components inhibiting NGS workflow
High throughput compatible—easy to automate on the Thermo Scientific KingFisher magnetic particle processors for high-throughput sample handling

Applications
DNA fragment library cleanup and size-selection for downstream use in next generation sequencing workflows.

Includes
• MagJET Magnetic Beads
• Binding Solution
• Wash Buffer (concentrated)
• Elution Buffer
• DNA Fragment Mix

TRUSTED_SUSTAINABILITY

Specifications

Format Kit
Isolation Technology Magnetic Bead
Sequencing Type Genome and DNA Sequencing
Final Product Type DNA
For Use With (Equipment) Magnetic Stand, KingFisher™ mL, KingFisher™ Flex
Product Line MagJET
Product Type NGS Cleanup and Size Selection Kit
Quantity 4 x 96 Preps
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.