Learn More
Thermo Scientific™ Reagents for MagJET™ Plant gDNA Kit
Catalog No. FERR2502 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
60 mL
Catalog No. Quantity
FERR2502 60 mL
1 options
Catalog No. FERR2502 Supplier Thermo Scientific™ Supplier No. R2502
Precipitate the sample lysate in order to isolate plant genomic DNA. Thermo Scientific™ MagJET™ Precipitation Solution is a component of the MagJET™ Plant gDNA Kits (K2761/2762) and may be purchased separately in 60mL aliquots.
The Thermo Scientific™ MagJET™ Plant Genomic DNA Kit is designed for fast and efficient purification of genomic DNA from a variety of plant species and tissues without phenol/chloroform extraction. The kit utilizes magnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) MagJET Plant gDNA Kit
Quantity 60 mL