Learn More
Thermo Scientific™ Elution Buffer for MagJET Plasmid DNA Kit
Catalog No. FERR2631 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
18 mL
Catalog No. Quantity
FERR2631 18 mL
1 options
Catalog No. FERR2631 Supplier Thermo Scientific™ Supplier No. R2631
The Thermo Scientific™ MagJET™ Plasmid DNA Kit is designed for fast and efficient purification of plasmid DNA from overnight cultures of E. coli cultures.
The Thermo Scientific™ MagJET™ Plasmid DNA Kit is designed for fast and efficient purification of plasmid DNA from overnight cultures of E. coli cultures. The kit utilizes magnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.
TRUSTED_SUSTAINABILITY
Specifications
Buffer Elution Buffer
Product Line MagJET
Quantity 18 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.