Learn More
Thermo Scientific™ Reagents for MagJET™ Plasmid DNA Kit
Catalog No. FERR2632 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
45 mL
Catalog No. Quantity
FERR2632 45 mL
1 options
Catalog No. FERR2632 Supplier Thermo Scientific™ Supplier No. R2632
Elute purified plasmid DNA for use in common molecular biology procedures. Thermo Scientific™ MagJET™ Elution Buffer is a component of the MagJET™ Plasmid DNA Kits (K2791/2792) and may be purchased separately when additional solution is needed. It is provided in 45mL aliquots.
The Thermo Scientific™ MagJET™ Plasmid DNA Kit is designed for fast and efficient purification of plasmid DNA from overnight cultures of E. coli cultures. The kit utilizes magnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) MagJET Plasmid DNA Kit
Quantity 45 mL