Learn More
Thermo Scientific™ FirstDefender™ RM Handheld Chemical Identification
Description
- Fast, accurate identification based on Raman spectroscopy to quickly identify unknown solid and liquid chemicals down range
- Sophisticated chemometric algorithms automatically determine presence of mixed chemicals
- The easy-to-navigate and extensive substance library includes explosives, toxic industrial chemicals (TICs), chemical warfare agents (CWAs), narcotics, precursors, white powders and more
- Lightweight, ergonomic design weighs 1.8 lb. (800g) for easy transportation down range
- Point-and-shoot™ identification operates directly through sealed translucent glass or plastic containers, avoiding exposure to potentially harmful substances; an integrated vial holder offers an alternative sampling method
- Easy to use and maintain with a menu driven user interface that requires minimal training and no scheduled maintenance or calibration
- Nondestructive testing—optical technique maintains sample integrity
- Can be powered using the included rechargeable battery, wall adapter, or SureFire™ disposable batteries
Certifications
Certified to MIL-STD 810G standards for ruggedness and environmental protection to IP67 standards against dust and water up to 1m.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Includes | 1-year warranty and support |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.