Learn More
Thermo Scientific™ FirstDefender™ RM Handheld Chemical Identification
GSA_VA
Catalog No. 17720413 Shop All Thermo Scientific Products
Change view
Click to view available options
Includes:
1-year warranty and support
2-year warranty and support
3-year warranty and support
4-year warranty and support
5-year warranty and support
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
17-720-413 5-year warranty and support
17-720-411 3-year warranty and support
17-720-409 1-year warranty and support
17-720-412 4-year warranty and support
17-720-410 2-year warranty and support
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-720-413 Supplier Thermo Scientific™ Supplier No. 8000017501
Add to Cart
Add to Cart
Request a Quote
Enhanced speed, performance, mixture analysis, and user interface make this Raman ideal for rapid, accurate identification of unknown chemicals directly in the field
  • Fast, accurate identification based on Raman spectroscopy to quickly identify unknown solid and liquid chemicals down range
  • Sophisticated chemometric algorithms automatically determine presence of mixed chemicals
  • The easy-to-navigate and extensive substance library includes explosives, toxic industrial chemicals (TICs), chemical warfare agents (CWAs), narcotics, precursors, white powders and more
  • Lightweight, ergonomic design weighs 1.8 lb. (800g) for easy transportation down range
  • Point-and-shoot™ identification operates directly through sealed translucent glass or plastic containers, avoiding exposure to potentially harmful substances; an integrated vial holder offers an alternative sampling method
  • Easy to use and maintain with a menu driven user interface that requires minimal training and no scheduled maintenance or calibration
  • Nondestructive testing—optical technique maintains sample integrity
  • Can be powered using the included rechargeable battery, wall adapter, or SureFire™ disposable batteries
Certifications
Certified to MIL-STD 810G standards for ruggedness and environmental protection to IP67 standards against dust and water up to 1m.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Includes 5-year warranty and support