Learn More
Thermo Scientific™ FirstDefender™ RMX Handheld Chemical Identification
GSA_VA
Catalog No. 17720418 Shop All Thermo Scientific Products
Change view
Click to view available options
Includes:
1-year warranty and support
2-year warranty and support
3-year warranty and support
4-year warranty and support
5-year warranty and support
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
17-720-418 5-year warranty and support
17-720-415 2-year warranty and support
17-720-414 1-year warranty and support
17-720-416 3-year warranty and support
17-720-417 4-year warranty and support
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-720-418 Supplier Thermo Scientific™ Supplier No. 8000021501
Add to Cart
Add to Cart
Request a Quote
Designed for flexible use modes; uses Raman spectroscopy to quickly identify unknown solid and liquid chemicals down range
  • Sophisticated chemometric algorithms automatically determine presence of mixed and contaminated chemicals
  • Flexible modes—offers handheld use with fixed probe or vial, or easily connects to a tactical robot through integrated RS232 port for universal control
  • Designed for use down range with a tough form factor, quick decontamination and easy operation in bulky protective gear
  • Lightweight, ergonomically designed unit weighs 2 lb. (919g) for easy transportation down range
  • Point-and-shoot sampling offers improved operation through sealed translucent containers; avoids exposure and maintains evidence
  • Noncontact analysis requires no consumables and no calibration
  • The easy-to-navigate vast substance library includes explosives, toxic industrial chemicals (TICs), chemical warfare agents (CWAs), narcotics, precursors, white powders and more
  • Easy to use and maintain with a menu driven user interface that requires minimal training and no scheduled maintenance or calibration
  • Can be powered using the included rechargeable battery, wall adapter or SureFire™ disposable batteries
Certifications
MIL-STD 810G and IP67 certified.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Includes 5-year warranty and support