Learn More
Thermo Scientific™ FirstDefender™ RMX Handheld Chemical Identification
Description
- Sophisticated chemometric algorithms automatically determine presence of mixed and contaminated chemicals
- Flexible modes—offers handheld use with fixed probe or vial, or easily connects to a tactical robot through integrated RS232 port for universal control
- Designed for use down range with a tough form factor, quick decontamination and easy operation in bulky protective gear
- Lightweight, ergonomically designed unit weighs 2 lb. (919g) for easy transportation down range
- Point-and-shoot sampling offers improved operation through sealed translucent containers; avoids exposure and maintains evidence
- Noncontact analysis requires no consumables and no calibration
- The easy-to-navigate vast substance library includes explosives, toxic industrial chemicals (TICs), chemical warfare agents (CWAs), narcotics, precursors, white powders and more
- Easy to use and maintain with a menu driven user interface that requires minimal training and no scheduled maintenance or calibration
- Can be powered using the included rechargeable battery, wall adapter or SureFire™ disposable batteries
Certifications
MIL-STD 810G and IP67 certified.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Includes | 3-year warranty and support |