Learn More
Fisherbrand™ Flexaframe™ Foot Plate
Catalog No. 1466625Q Shop All Fisherbrand Products
Change view
Click to view available options
Material:
Metal
Catalog No. Material
14-666-25Q Metal
1 options
Catalog No. 14-666-25Q
Mounts lattice permanently
  • Foot plate holds Flexaframe poles
  • Secures them to floors, walls and tables with screws
  • Made of Castaloy™ metal
Specifications
Material Metal
Product Type Foot Plate
For Use With (Application) Mounts lattice permanently, secures poles to floors, walls and tables with screws